PurFlock™ Ultra Sterile Flocked Swabs
Actual product may vary
Puritan
PurFlock™ Ultra Sterile Flocked Swabs
Packaged in a reclosable dry transport tube for ease in transport to the lab
Have Questions?
Catalog NumberLength (Metric)Tip StyleIncludes
FIS22-029-614153.4 mmMinitip flockedBreakpoint length: None
Catalog number FIS22-029-614
Price (SGD)
1,227.00
Case
Length (Metric):
153.4 mm
Tip Style:
Minitip flocked
Includes:
Breakpoint length: None
Price (SGD)
1,227.00
Case
Packaged in a reclosable dry transport tube for ease in transport to the lab
  • Medical grade quality
  • Polystyrene handle with or without molded breakpoint (at 30, 80, 100mm)
  • Highly absorbent, soft smooth tip
  • Available in Mini/Standard/Large/Elongated/Ultrafine/Micro ultrafine tips
  • Shelf life is three years
Dry Transport System
  • Made from virgin polypropylene resin
  • Securely holds swab in round bottom tube
  • Creates safe and sterile environment to accommodate many different styles of collection devices
  • Injection molded plug-style cap assures precision fit for multiple handle styles
  • Distinctive label on tubes for describing product identity, expiry date, patient identification and collection information
Buccal cell collection, forensic evidence collection, nasopharyngeal and urethral cell collection, DNA collection, rapid diagnostic testing, DNA collection and tesing, collection of bacteria or microorganisms
Specifications
Quantity500/Cs.
Certifications/ComplianceCE, ISO 9001:2008, ISO 13485:2003
Length (English)6.04 in.
Product TypeFlocked Swab
Length (Metric)153.4 mm
Length (English) Tip0.725 in.
Length (Metric) Tip18.41 mm
Diameter (Metric) Tip3.42 mm
Diameter (English) Tip0.135 in.
IncludesBreakpoint length: None
PackagingDry transport tube
Shelf Life3 Year(s)
SterilitySterile
Handle MaterialPolystyrene
Tip StyleMinitip flocked
Unit SizeCase