Calgiswab™ Calcium Alginate Tipped Applicator
Actual product may vary
Puritan
Calgiswab™ Calcium Alginate Tipped Applicator
Calcium alginate tip contains no fatty acids and is ideal for specimen collection
Have Questions?
Catalog NumberHandle Material
FIS22-029-554Wood
Catalog number FIS22-029-554
Price (SGD)
112.00
Pack
Handle Material:
Wood
Price (SGD)
112.00
Pack
  • General all purpose swab for wound care
  • Wooden or semi-flexible polystyrene handle
  • Sterile
  • Medical grade quality
  • Individually wrapped
Specimen collection, wound care
Specifications
Certifications/ComplianceCE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Length (English)6 in.
For Use With (Application)For use in specimen collection and wound care
Product TypeCalgiswab™ Calcium Alginate Tipped Applicator
Length (Metric)152.4 mm
Length (English) Tip0.64 in.
Length (Metric) Tip16.2 mm
Diameter (Metric) Tip5.5 mm
Diameter (English) Tip0.219 in.
GradeMedical
PackagingIndividually-wrapped
SterilitySterile
Handle MaterialWood
Unit SizePack