Cotton-Tipped Applicators
Actual product may vary
Puritan
Cotton-Tipped Applicators
For specimen collection, testing and screening
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTip StyleHandle MaterialLength (Metric) HandleSterility
FIS22-029-553CottonWood14.7 cmSterile
FIS19-062-616RegularWood14.7 cmNon-sterile
Catalog number FIS22-029-553
Price (SGD)
383.00
Case
Tip Style:
Cotton
Handle Material:
Wood
Length (Metric) Handle:
14.7 cm
Sterility:
Sterile
Price (SGD)
383.00
Case
Various style available: sterile/non-sterile, wood/paper handles, tip styles and length.
Specifications
Certifications/ComplianceCE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Diameter (English) Handle0.1 in.
Length (English)6 in.
For Use With (Application)Specimen collection, wound dressing and cleaning, cell collection for DNA Testing, cosmetic applications
Product TypeApplicator
Diameter (Metric) Handle0.254 cm
Length (Metric)15.2 cm
Length (English) Handle5.8 in.
Length (Metric) Handle14.7 cm
Length (English) Tip0.625 in.
Length (Metric) Tip1.5 cm
Diameter (Metric) Tip0.47 cm
Diameter (English) Tip0.188 in.
GradeMedical
SterilitySterile
Handle MaterialWood
Tip StyleCotton
Tip MaterialCotton
Unit SizeCase