Specifications
DescriptionHorizontal Centrifuge, Timer, Up to 10 mL Tubes
IncludesRotor, Tube Holders
TypeCentrifuge, Fixed Angle
Capacity6 x 3 to 10 mL
Max. RCF1,600 x g
Max. Speed3,380 rpm
RefrigeratedNo
Profile (Acceleration/Braking)Acceleration: 15 sec., Deceleration: 80 sec.
Electrical Requirements115 V ±10, 60 Hz
Warranty2 Years
Product Line600 Series
DisplayColor Graphic Display
Height (English) Exterior8.75 in.
Height (Metric) Exterior22 cm
Length (English) Exterior14 in.
Length (Metric) Exterior36 cm
Safety FeaturesPositive Lid Latching, Clear Shatter Proof Lid, Reinforced Side Walls
TimerMechanical
Width (English) Exterior11.75 in.
Width (Metric) Exterior30 cm
Rotor6-place Horizontal
Wattage165 W
Controller TypeTimer
VentilatedYes
Weight (English)11 lb.
Weight (Metric)5 kg
For Use With (Application)Clinical
Quantity1 Ea.
Maximum Force Range1,000 to 1,999 x g
Maximum Speed Range3,000 to 3,999 rpm
Rotor IncludedYes
Dimensions (HWD)8.75 x 11.75 x 14 in. (22.2 x 29.8 x 35.6 cm)
Drive SystemAC
Net Weight11 lb. (5 kg)
Run Time1 to 30 min.
Frequency50/60 Hz
Voltage115 V
Unit SizeEach