▸
First Aid and Medical
▸
Patient Care Products
▸
Medical Syringes
▸
SUN-SRi™ Luer-Slip Syringes
Actual product may vary
Thermo Scientific
SUN-SRi™ Luer-Slip Syringes
Siliconized and assembled with scale and tip shield.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Product Type
Sterility
FIS22-028853
1 mL
Luer-slip Syringe
Sterile
FIS22-314383
2 mL
Luer-slip Syringe
Non-sterile
2 Options
Catalog number
FIS22-028853
Price (SGD)
-
Volume (Metric):
1 mL
Product Type:
Luer-slip Syringe
Sterility:
Sterile
Syringe, Chemically resistant, inert polypropylene and polyethylene, Luer Slip, Bulk
Chemically resistant, inert polypropylene and polyethylene
No rubber plunger seals or silicone lubricants that may cause sample contamination
Bulk packaged, choice or sterile or nonsterile
LUER-Slip connection
Specifications
Material
Plastic
Product Type
Luer-slip Syringe
Disposable
Single-use
Graduations
0.01 mL
Volume (Metric)
1 mL
Needle Point Style
Centered
Connection Type
Leur Slip
Sterility
Sterile