Scintillation Vial Rack
Actual product may vary
Fisherbrand
Scintillation Vial Rack
For start to finish sample preparation
Have Questions?
Catalog NumberHolds
FIS22-028498Vials
Catalog number FIS22-028498
Price (SGD)
124.00
Each
Holds:
Vials
Price (SGD)
124.00
Each
  • Holds 24 scintillation vials of 30 mm diameter for storage, handling and sample preparation
  • Rugged, stable design molded from corrosion-resistant polypropylene
  • Autoclavable
  • Shelf height 4.4 cm (1.75 in.)
Specifications
AutoclavableAutoclavable
ColorWhite
HoldsVials
MaterialPolypropylene
Positions24
ShapeRectangular
StackableNo
SterilityNon-sterile
For Use With (Equipment)30mm dia. Scintillation vials
QuantityPk. of 1
Dimensions (L x W x H)11.75 x 5 x 3.75 in. (29.8 x 12.7 x 9.5cm)
Length (Metric)29.8 cm
Length (English)11.75 in.
Width (English)5 in.
Height (English)3.75 in.
Product TypeVial Rack
Diameter (Metric) Inner30 mm
Shelves2 Tiers
Height (Metric)9.5 cm
Array8 x 3
Width (Metric)12.7 cm
Unit SizeEach