Finntip™ Extended Length Pipette Tips
Actual product may vary
Finntip™ Extended Length Pipette Tips
Finntip™ Extended Length Pipette Tips
Thermo Scientific
Finntip™ Extended Length Pipette Tips
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FormatSterility
FIS22-028146200 μLRackNon-sterile
9400100200 μLBoxNon-sterile
FIS21-377-43200 μLBoxNon-sterile
9400130200 μLRackNon-sterile
94014101000 μLBulkNon-sterile
94014201000 μLRackNon-sterile
94014231000 μLRackSterile
FIS21-377-6011000 μLRackSterile
Catalog number FIS22-028146
Price (SGD)
552.00
Case
Volume (Metric):
200 μL
Format:
Rack
Sterility:
Non-sterile
Price (SGD)
552.00
Case
Ensure accurate, precise pipetting. Thermo Scientific™ Finntip™ Pipette Tips are manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips, to prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked or bulk
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Tip Range
  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 200 Ext, Non-Filtered, Non-sterile, Rack
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ModelFinntip 200 Ext
Length (English)3 in.
Length (Metric)7.8cm
FormatRack
Color CodeYellow
For Use With (Equipment)Thermo Scientific Finnpipette 200
Product LineFinnTip
Fit for PurposeCertified Quality
Unit SizeCase