SPECI-GARD™ Specimen Biohazard Transport Bags
SPECI-GARD™ Specimen Biohazard Transport Bags
Actual product may vary
Minigrip
SPECI-GARD™ Specimen Biohazard Transport Bags
Used for specimen transport in hospitals and labs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Includes
FIS22-02675317 x 15 in.2 Pockets
FIS22-31425710 x 6 in.2 Pockets
FIS22-3079359 x 5 in.2 Pockets
FIS22-307-94810 x 10 in.2 Pockets, Instructions
FIS22-310-0946 x 6 in.1 Pocket
FIS22-310-00110 x 6 in.2 Pockets, Red and Black Print
FIS22-310-00410 x 6 in.Instructions
FIS22-310-0959 x 6 in.2 Pockets, 3 x 3 in. Absorbent Sheet
Catalog number FIS22-026753
Price (SGD)
-
Dimensions (L x W):
17 x 15 in.
Includes:
2 Pockets
  • Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
  • With printed biohazard logo
  • Separate outside pocket holds the requisition slip
  • Traditional scored opening
  • No glove-to-adhesive contact, ensuring ease of use
  • Once closed, the bag stays closed, even in pneumatic tube systems
  • Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
  • Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
  • To open, simply snap at score
  • Disposable, eliminating the possibility of cross-contamination caused by reusing bags
  • Manufactured with Micro-Tex™ polyethylene for slip-free handling
Specifications
Closure TypePress and Close Liquid Tight
ColorClear
DisposableSingle-use
For Use With (Application)Used for specimen transport in hospitals and labs
Includes2 Pockets
MaterialPolyethylene
Dimensions (L x W)17 x 15 in.
Length (English)17 in.
Length (Metric)43.18 cm
Width (English)15 in.
Width (Metric)38.1 cm
Height (English)17 in.
Height (Metric)43.18 cm
Product TypeBiohazard Specimen Transport Bag
Unit SizeCS