Check Order Status
Wheaton™ V Vial™ with Crimp Top Finish; no Closures
Actual product may vary
DWK Life Sciences
Wheaton™ V Vial™ with Crimp Top Finish; no Closures
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Height (Metric)
Diameter (Metric) Outer
Volume (English)
FIS22-021832
5 mL
61 mm
20 mm
1.4 dr.
FIS03-410-036
3 mL
46 mm
20 mm
0.8 dr.
2 Options
Catalog number
FIS22-021832
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5 mL
Height (Metric):
61 mm
Diameter (Metric) Outer:
20 mm
Volume (English):
1.4 dr.
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
Not graduated
Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
Conical interior bottoms are designed to provide total downward drainage for maximum sample retrieval
Specifications
Diameter (English) Inner
0.51 in.
Diameter (Metric) Inner
13 mm
Color
Clear
Diameter (Metric) Outer
20 mm
Material
Borosilicate Glass (Low Extractable)
Volume (Metric)
5 mL
For Use With (Application)
Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Graduated
No
Style
Conical V
Volume (English)
1.4 dr.
Closure Type
Crimp Cap
Height (Metric)
61 mm
Certifications/Compliance
ASTM E438, USP Type I, Type I, Class A