Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Racks
▸
Freezer Storage Racks
▸
Workstation Rack for CryoClear™ Vials
Actual product may vary
Globe Scientific Inc
Workstation Rack for CryoClear™ Vials
Accommodates both round bottom and self-standing CryoClear™ vials, cryogenic vials and sample tubes.
Have Questions?
Catalog Number
Color
FIS22-010-1189
Blue
Catalog number
FIS22-010-1189
Price (SGD)
84.00
Each
Add to cart
Color:
Blue
Price (SGD)
84.00
Each
Add to cart
Format: 10 x 4 (40-place)
Material: Polycarbonate (PC)
Anti-slip rubber feet
Autoclavable at 121°C, 15 psig (1 bar) for 20 min.
Temperature range: -196°C to +121°C
Dimensions: 215 x 100 x 25mm
Specifications
Color
Blue
Holds
40 Vials
Array
10 x 4
Material
Polycarbonate
Product Type
Workstation Rack