Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits
Actual product may vary
Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits
Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits
Smiths Medical
Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits
Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results.The syringes are available with a broad choice of product features.
Have Questions?
Catalog NumberQuantityProduct TypeNeedle Gauge
FIS22-004-185200/Cs.Filter-Pro, Needle-Pro Device25 G
Catalog number FIS22-004-185
Price (SGD)
1,205.00
Case
Quantity:
200/Cs.
Product Type:
Filter-Pro, Needle-Pro Device
Needle Gauge:
25 G
Price (SGD)
1,205.00
Case
  • Advanced-formula dry lithium heparin
  • Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
  • Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
  • Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
  • Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
  • Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
  • Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Specifications
Needle Gauge25 G
For Use With (Application)Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Includes1mL luer-slip syringe with Filter-Pro™ device, and 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant
Product TypeFilter-Pro, Needle-Pro Device
Quantity200/Cs.
PackagingSterile Tyvek™
Shelf Life3 Years
Volume (Metric)1 mL