Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Racks
▸
Pipet Racks
▸
Extra Wide Countertop Serological Pipet Holder
Actual product may vary
Poltex
Extra Wide Countertop Serological Pipet Holder
Holder keeps pipets tidy and convenient when used on countertop
Have Questions?
Catalog Number
Mounting
FIS22-000-188
Countertop
Catalog number
FIS22-000-188
Price (SGD)
485.00
Each
Add to cart
Mounting:
Countertop
Price (SGD)
485.00
Each
Add to cart
Shelves are removable for easy cleaning
Slanted top allows for extra storage of other items
Pocket dimensions:
13.97cm (5.5 in.) wide
27.94cm (11 in.) deep
5.71, 6.98, 8.25, 10.79cm (2.25, 2.75, 3.25 and 4.25 in.) tall
Specifications
For Use With (Equipment)
Serological Pipets
Mounting
Countertop
Dimensions (L x W x H)
28.39 x 15.25 x 40.64cm (11.18 x 6 x 16 in.)
Product Type
Pipet Holder
Shelves
4 Tiers