Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Eye Protection and Face Protection
▸
Benchtop Splash Shields
▸
Articulating Biohazard Shield
Actual product may vary
Poltex
Articulating Biohazard Shield
Provides full range of motion to protect from biohazard splash
Have Questions?
Catalog Number
Product Type
FIS22-000-185
Two articulated stiff stems
Catalog number
FIS22-000-185
Price (SGD)
255.00
Each
Add to cart
Product Type:
Two articulated stiff stems
Price (SGD)
255.00
Each
Add to cart
Constructed of break resistant polycarbonate (3/16 base, 1/8 shield)
Comes in two versions: 6.75 in.× 6.75 in. with one stem, and 11.75 in.×11.75 in. with two stems
Articulating stem allows maximum range of motion to protect the user from biohazard splash
Shield and base units are 6.75 in.× 6.75 in. or 11.75 in.×11.75 in.
Rubber bumpons add a non-slip base, and a stiff stem holds the shield in place
Specifications
Product Type
Two articulated stiff stems
Material
Polycarbonate