Safety Lancet
Actual product may vary
MediPurpose
Safety Lancet
Safety needle or blade lancet is safely concealed before and after use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorIncludesNeedle Gauge
FIS22-000-10375 to 100μLGreenBlade19g
FIS22-000-10120 to 40μLOrangeNeedle22g
Catalog number FIS22-000-103
Price (SGD)
-
Capacity (Metric):
75 to 100μL
Color:
Green
Includes:
Blade
Needle Gauge:
19g
  • Unique, one-step activation mechanism for fingerstick sampling—no arming is required
  • Once lancet is used, it is rendered inoperative, providing added safety for patient and clinician
  • High-speed delivery and penetration method minimizes patient pain and clinician error
  • Latex-free
  • Available with either a needle or blade, each with different depths of penetration, to provide optimal blood flow for different applications and skin types
Specifications
Capacity0.0025 to 0.00338 oz., 75 to 100μL
Capacity (English)0.0025 to 0.00338 oz.
Capacity (Metric)75 to 100μL
Certifications/ComplianceOSHA
ColorGreen
Depth (English)0.7 in.
Depth (Metric)1.8mm
For Use With (Application)For all POC tests requiring whole Blood samples, including Glucose, cholesterol, HIV and more
IncludesBlade
Needle Gauge19g
Quantity100 Pack
Product TypeSafety Lancet
Unit SizePack