Reagent Reservoirs: 12-Channel
Actual product may vary
VistaLab Technologies
Reagent Reservoirs: 12-Channel
Unique trough-within-trough design maximizes reagent recovery.
Have Questions?
Catalog NumberSterilityPackagingQuantity
FIS21-385-120SterileIndividually Wrapped10/Cs.
Catalog number FIS21-385-120
Price (SGD)
190.00
Case
Sterility:
Sterile
Packaging:
Individually Wrapped
Quantity:
10/Cs.
Price (SGD)
190.00
Case
  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • 100mL model (Cat. No. 21-382-111) supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
  • 5 mL capacity / well
Specifications
Certifications/ComplianceRNase/DNase, Pyrogen-free
ColorClear
Capacity (Metric)60 mL
MaterialPolypropylene
SterilitySterile
TypeReagent Reservoir
For Use With (Application)General Liquid Handling
Height (English)4 in.
IncludesLid
Length (English)11 in.
PackagingIndividually Wrapped
Quantity10/Cs.
Volume (Metric) Well5 mL
Width (English)6 in.
AutoclavableAutoclavable
Channels12
DisposableSingle-use
Bottom ShapeV-bottom
Unit SizeCase