Xenin 8
Xenin 8
Actual product may vary
Xenin 8
Xenin 8
Xenin 8
Neurotensin-like peptide; potentiates insulin and glucagon secretion
Have Questions?
Catalog NumberQuantity
FIS21-3811 mg
Catalog number FIS21-381
Price (SGD)
624.00
PK
Add to cart
Quantity:
1 mg
Price (SGD)
624.00
PK
Add to cart
Xenin 8 is a C-Terminal fragment of xenin, a neurotensin-like peptide; modulates pancreatic insulin and glucagon secretion/effects. Stimulates basal and arginine-induced insulin secretion and potentiates the insulin response to glucose (EC50 = 0.16 nM). Also potentiates arginine- and carbachol-induced glucagon secretion in a somatostatin-independent manner.
For Research Use Only
Specifications
DescriptionXenin 8
Molecular Weight (g/mol)1046.28
Product TypeXenin 8
Quantity1 mg
SequenceHis-Pro-Lys-Arg-Pro-Trp-Ile-Leu
Unit SizePK