MLA™ Brand Digital Pipetters
MLA™ Brand Digital Pipetters
Actual product may vary
VistaLab Technologies
MLA™ Brand Digital Pipetters
Perfect for tests that require multiple or nonstandard volumes
Have Questions?
Catalog NumberVolume (Metric)AccuracyColorCompatible Tips
FIS21-369H200 to 1000 μL±1%Gray1mL Tip
Catalog number FIS21-369H
Price (SGD)
2,307.00
EA
Add to cart
Volume (Metric):
200 to 1000 μL
Accuracy:
±1%
Color:
Gray
Compatible Tips:
1mL Tip
Price (SGD)
2,307.00
EA
Add to cart
Three adjustable models cover volume range: 10 to 1000μL. To adjust volume, simply turn volume selector knob near top of pipetter. Four-digit reading is locked in by click stops, ensuring accuracy.
  • One-handed operation
  • D-Tipper™ feature allows you to pick up and dispose of tips without touching them
  • Volume range conveniently marked on all models
  • Factory calibrated for accurate delivery
  • Easy in-lab recalibration
  • Made of lightweight aluminum and stainless steel to assure durability and long life
Specifications
Accuracy±1%
Compatible Tips1mL Tip
AutoclavableNot Autoclavable
ColorGray
DisposableReusable
For Use With1 mL Tip
Grip TypeErgonomic
MaterialAluminum, Stainless Steel
Number of Channels1
Product LineMLA Brand
ProgrammabilityNo
SterilityNon-sterile
TypeDigital Pipetter
Volume (Metric)200 to 1000 μL
For Use With (Application)Multiple or Non-Standard Volumes
For Use With (Equipment)1 mL Tip
IncludesNozzle Insert, Calibration Key
Quantity1 Ea.
WarrantyLimited Lifetime
Length (English)9 in.
Width (English)2 in.
Height (English)1.5 in.
Unit SizeEA