ART™ Wide Bore Filtered Pipette Tips
Actual product may vary
Thermo Scientific
ART™ Wide Bore Filtered Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Catalog NumberVolume (Metric)Fit for Purpose
FIS21-236-2C1000 μLSterile and PCR Ready
Catalog number FIS21-236-2C
Price (SGD)
446.00
Pack
Volume (Metric):
1000 μL
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
446.00
Pack
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
DescriptionART 1000G, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ModelART 1000G
Length (English)3.36 in.
Length (Metric)8.54cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatLift-off Lid Rack
Tip StyleGenomic/Wide Orifice
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePack