Snap-N-Rack Tube Racks: 24-Place
Actual product may vary
Globe Scientific Inc
Snap-N-Rack Tube Racks: 24-Place
Shipped flat to reduce shelf space and easily snaps together for use
Have Questions?
Catalog NumberColor
FIS21-200-281White
Catalog number FIS21-200-281
Price (SGD)
87.00
Each
Color:
White
Price (SGD)
87.00
Each
For use with 30mm tubes
  • Made of Polypropylene
  • Freezable, without deforming or going brittle
  • Non-floating in water baths
  • Alphanumerical markings are molded into racks for easy test tube identification
  • Assorted colors available for sample coding
  • Autoclavable at 121°C for 20 minutes
Specifications
ColorWhite
HoldsTubes (30 mm)
MaterialPolypropylene
Positions24
Dimensions (L x W x H)11.61 x 4.41 x 3.34 in. (30 x 11.2 x 8.5 cm)
Length (Metric)30 cm
Length (English)11.61 in.
Width (English)4.41 in.
Height (English)3.34 in.
Diameter (Metric) Opening30 mm
Product TypeTube Rack
Height (Metric)8.5 cm
Width (Metric)11.2 cm
Unit SizeEach