Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Spectroscopy
▸
Spectrophotometry
▸
Spectrophotometer Sample and Cell Holders
▸
Accessories for Spectrophotometer Cuvettes
Actual product may vary
Globe Scientific Inc
Accessories for Spectrophotometer Cuvettes
For use with Spectophotometer cuvettes
Have Questions?
Catalog Number
Material
Type
FIS21-200-262
Polystyrene
Stirring Stick
Catalog number
FIS21-200-262
Price (SGD)
40.00
Pack
Add to cart
Material:
Polystyrene
Type:
Stirring Stick
Price (SGD)
40.00
Pack
Add to cart
Polyethylene made 12-place cuvette holder of 170 x 50 x 15mm dimensions
LDPE snap caps for square cuvettes for added protection against spillage and contamination
Polystyrene disposable 90mm stirring stick for access to the bottom of cuvettes
Specifications
Type
Stirring Stick
Disposable
Single-use
For Use With (Equipment)
Spectrophotometer cuvettes
Length (English)
3.54 in.
Length (Metric)
90 mm
Material
Polystyrene