Sedi-Rate™ ESR System Starter Kit
Actual product may vary
Sedi-Rate™ ESR System Starter Kit
For Westergren ESR determination
Have Questions?
Catalog NumberQuantity
FIS21-200-213100 Tests
Catalog number FIS21-200-213
Price (SGD)
-
Quantity:
100 Tests
  • Closed system for optimum safety
  • Produces accurate and reproducible results
  • Supplied with Polystyrene, graduated pipet with fibrous plug located at zero mark
  • Contains Polypropylene vial pre-filled with 0.25mL of 3.8% trisodium citrate diluent
  • Eliminates splashing, spraying and aerosol hazards
Specifications
TypeSedi-Rate™ Westergren Starter Kit
For Use With (Application)Westergren ESR determination
Includes100 Sedi-Rate™ Polystyrene graduated pipettes with fibrous plug, Polypropylene vial pre-filled with 0.25mL of 3.8% Trisodium Citrate diluent, 10-place rack with pipette support clamps, leveling platform, 100 piercing funnels and 100 ESR
Quantity100 Tests
Unit SizePack