Check Order Status
PCR Micropipets
Actual product may vary
Drummond
PCR Micropipets
Disposable, positive displacement micropipets with stainless steel plungers
Have Questions?
Change view
Catalog Number
Volume (Metric)
FIS21-179A
5 μL
FIS21-179B
1 to 10 μL
2 Options
Catalog number
FIS21-179A
Price (SGD)
284.00
PK
Add to cart
Volume (Metric):
5 μL
Price (SGD)
284.00
PK
Add to cart
Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
Decontaminate at temperature of 400°F or higher
Specifications
For Use With (Application)
To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
Includes
Stainless Steel plungers
Volume (Metric)
5 μL
Type
PCR Micropipet
Disposable
Single-use