Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes
Actual product may vary
Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes
Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes
Drummond
Hemato-Clad™ Mylar™-Wrapped Hematocrit Tubes
Combine the precision of glass with the safety of Mylar wrapping
Have Questions?
Catalog NumberAdditiveClosure Color
FIS21-176-5NoneBlue
Catalog number FIS21-176-5
Price (SGD)
42.00
Pack
Additive:
None
Closure Color:
Blue
Price (SGD)
42.00
Pack
Combine the precision of glass with the safety of Mylar wrapping
  • The chance of contamination due to glass fragments or aerosols is greatly reduced
  • Accidental breakage is significantly reduced, particularly when pressing into clay sealant (not included)
  • In the event of breakage, the Mylar wrapping will help contain the blood sample and provide an added layer of protection against cuts from broken glass
  • Available both plain and heparinized
Specifications
Product TypeMylar™ Wrapped Hematocrit Tube, Plain
AdditiveNone
MaterialGlass
Diameter (Metric) Inner0.11 to 0.12 cm
Closure ColorBlue
For Use With (Application)For Blood collection
Length (Metric)7.5 cm
Length (English)2.95 in.
Certifications/ComplianceFDA, NIOSH, CDC, OSHA
Diameter (English) Inner0.04 to 0.047 in.
Unit SizePack