Short-Length Microcaps™ Micropipets
Short-Length Microcaps™ Micropipets
Actual product may vary
Drummond
Short-Length Microcaps™ Micropipets
Ideal for handling dangerous materials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)Diameter (Metric) InnerDiameter (Metric) Outer
FIS21-170H25 μL65 mm0.7 mm0.97 mm
FIS21-170E5 μL32 mm0.44 mm0.93 mm
Catalog number FIS21-170H
Price (SGD)
44.00
PK
Add to cart
Volume (Metric):
25 μL
Length (Metric):
65 mm
Diameter (Metric) Inner:
0.7 mm
Diameter (Metric) Outer:
0.97 mm
Price (SGD)
44.00
PK
Add to cart
Ideal for handling dangerous materials
Precision-made capillary glass tubing, accurate to within ±1%.
  • To use, insert pipet through cap on end of rubber bulb assembly until visible through glass tube
  • Hole in rubber bulb permits free air circulation, fills pipet by capillary action
  • Pipet contains stated volume when completely filled from end to end
  • To discharge, hold finger over hole in bulb and squeeze
  • To rinse, repeat operation with diluent
Specifications
Accuracy±1%
CalibrationTo Contain
DescriptionNon-heparinized
Diameter (English) Inner0.0276 in.
Diameter (English) Outer0.0382 in.
Diameter (Metric) Inner0.7 mm
Diameter (Metric) Outer0.97 mm
MaterialGlass
For Use With (Application)For handling dangerous materials, fast, safe, clean and accurate way to transfer liquids in micro quantities
IncludesBulb assembly
Length (English)2.55 in.
Volume (Metric)25 μL
Length (Metric)65 mm
TypeMicrocaps™ Micropipet