Check Order Status
Replacement Bores for Drummond Micropipetters
Actual product may vary
Drummond
Replacement Bores for Drummond Micropipetters
Replacement parts
Have Questions?
Change view
Catalog Number
Type
FIS21-169B
Replacement Bore (20 μL)
FIS21-169A
Replacement Bore (10 μL)
2 Options
Catalog number
FIS21-169B
Price (SGD)
47.00
PK
Add to cart
Type:
Replacement Bore (20 μL)
Price (SGD)
47.00
PK
Add to cart
Replacement Parts
Specifications
Color
Black
Material
Glass
Type
Replacement Bore (20 μL)
For Use With (Equipment)
Drummond Micropipetter, digital microdispensers