▸
Protein Analysis Reagents
▸
Dialysis and Desalting
▸
General Dialysis Supplies
▸
Locking Dialysis Membrane Clamps
Locking Dialysis Membrane Clamps
Provides a leakproof seal.
Have Questions?
Catalog number
FIS21-153-100
Price (SGD)
-
Provides a leakproof seal.
Stay locked until released
Do not float
Constructed of Polyamide 6.6 from DuPont (NYLON)
Available in 2 sizes: 46mm or 65mm
10 clamps/pkg
Specifications
Product Type
Dialysis Membrane Clamp, up to 46 mm sealing width
Material
Polyamide 6.6 Dupont (Nylon)
Quantity
10 Pack
For Use With (Application)
Do not float, stay locked until released