Check Order Status
Kuehne Coverglass Thumb Forceps, 4 in. (10.2cm)
Actual product may vary
Thermo Scientific
Kuehne Coverglass Thumb Forceps, 4 in. (10.2cm)
Choose 4 in. (10.2cm) Kuehne Coverglass Thumb Forceps, featuring angled tips for quality and precision.
Have Questions?
Catalog Number
Product Type
FIS19060
Kuehne Coverglass
Catalog number
FIS19060
Price (SGD)
-
Product Type:
Kuehne Coverglass
Features:
Properly hardened, tempered and passivated
Electronically polished and cleaned for a high-quality finish
Patterns, dimensions and finish held to exacting standards
Specifications
Product Type
Kuehne Coverglass
Length (English)
4 in.
Length (Metric)
10.2 cm