Laboratory Storage Shelves
Actual product may vary
S-Curve
Laboratory Storage Shelves
Three separate shelves for storage
Have Questions?
Catalog NumberWidth (English)
FIS19-999-93024 in.
Catalog number FIS19-999-930
Price (SGD)
917.00
Each
Width (English):
24 in.
Price (SGD)
917.00
Each
  • 0.25 in.thick (0.6cm) clear Acrylic material
  • Can be wall mounted or used on the benchtop
  • Rubber pads on the bottom prevent slipping and scratching
  • Protective 2 in. (5cm) edge helps prevent spills and accidents
  • Flame-polished edges for a smooth finish
Specifications
ColorClear
MaterialAcrylic
Depth (English)8 in.
Depth (Metric)20.32 cm
Height (English)18 in.
Height (Metric)45.7 cm
Width (English)24 in.
Width (Metric)60.96 cm
Unit SizeEach