Smart Fabrication™ Work Tables with Enclosed Bases 5in. Backsplash
Actual product may vary
Advance Tabco
Smart Fabrication™ Work Tables with Enclosed Bases 5in. Backsplash
Advance Tabco Smart Fabrication™ Work Table with Enclosed Bases feature hinged doors for added convenience to the user.
Have Questions?
Catalog NumberDimensions (L x W x H)Weight (English)Weight (Metric)
FIS19-999-56248 x 24 x 35 in.270 lb.122.4 kg
Catalog number FIS19-999-562
Price (SGD)
14,879.00
Each
Dimensions (L x W x H):
48 x 24 x 35 in.
Weight (English):
270 lb.
Weight (Metric):
122.4 kg
Price (SGD)
14,879.00
Each
  • Available in two different width sizes: 24 in. (61cm), 30 in. (76.2cm)
  • 1.625 in. (4cm) sanitary rolled rim on the front side
  • 1.625 in. (4cm) square bend edges on the 2 sides
  • 5 in. (13cm) splash with 1 in. (2.54cm) return on rear side
  • Double panel hinged doors with recessed inner panels for extra rigidity
  • TIG welded
  • Exposed areas finished to match adjacent surfaces
  • Entire Top is mechanically polished to a satin finish
  • Hat section secured to Top reinforces and maintains level working surface
  • Gussets welded to support hat sections
  • Top is 14 gauge stainless steel type 304 series with galvanized understructure
  • Body is 18 gauge stainless steel type 430 series
  • Legs are 1.625 in. (4cm) diameter tubular stainless steel with stainless steel gussets and 1 in. (2.5cm) adjustable stainless steel bullet feet
Specifications
Dimensions (L x W x H)48 x 24 x 35 in.
Height (English)35 in.
Height (Metric)88.9 cm
IncludesEnclosed bases 5 in. backsplash
Length (English)48 in.
Length (Metric)122 cm
Weight (English)270 lb.
Weight (Metric)122.4 kg
Width (English)24 in.
Width (Metric)60.96 cm
Product TypeWorktable