Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Spill Control and Containment
▸
Secondary Containment
▸
HDPE, Basin
Actual product may vary
Justrite
HDPE, Basin
For Use With:
13.5L or 20L Carboys
Have Questions?
Catalog Number
Capacity (English)
FIS19-830-516
9 gal.
Catalog number
FIS19-830-516
Price (SGD)
235.00
Each
Add to cart
Capacity (English):
9 gal.
Price (SGD)
235.00
Each
Add to cart
Constructed of rugged high-density polyethylene
For use with 2.5, 4, 5, 10, 13.5, 20, 40 or 60L carboys
Specifications
Color
Red
Capacity (English)
9 gal.
Capacity (Metric)
34 L
Material
HDPE
Type
Basin
For Use With (Equipment)
13.5 L or 20 L Carboys
Dimensions (L x W x H)
19.5 x 15.5 x 10 in.