Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Carboys
▸
Carboy Spigots and Accessories
▸
Activated Carbon Filter (2 Pack)
Actual product may vary
Justrite
Activated Carbon Filter (2 Pack)
For Use With:
VaporTrap Solvent Waste Systems
Have Questions?
Catalog Number
No. per Pack
FIS19-830-506
2 Pk.
Catalog number
FIS19-830-506
Price (SGD)
727.00
Pack
Add to cart
No. per Pack:
2 Pk.
Price (SGD)
727.00
Pack
Add to cart
For use with VaporTrap™ solvent waste kits
Specifications
Description
Activated Carbon Filter
Diameter (English) Outer
1.9 in.
For Use With (Equipment)
VaporTrap Solvent Waste Systems
Length (English)
7 in.
Material
Charcoal-activated Carbon
No. per Pack
2 Pk.
Product Type
Filter