Tyvek™ 400 Frocks, Mandarin Collar
Actual product may vary
DuPont
Tyvek™ 400 Frocks, Mandarin Collar
Flash-spun high-density polyethylene provides an ideal balance of protection, durability, and comfort
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-823-600Medium
FIS19-823-601Large
FIS19-813-2155Small
FIS19-823-602X-Large
Catalog number FIS19-823-600
Price (SGD)
570.00
Case
Size:
Medium
Price (SGD)
570.00
Case
Ideal for lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
  • Inherent barrier to small size particles built into the fabric itself — no films or laminates to abrade or wear away
  • Body-length jacket with five front snap closures for easy donning and doffing
  • Enhanced frontal protection
  • Mandarin collar fits tightly around hooded PAPR
  • Elastic opening for tighter fit at wrist
  • Serged seams
  • White
Specifications
SizeMedium
MaterialTyvek
ColorWhite
DisposableLimited-use
Closure TypeSnaps
Cuff StyleElastic
Collar TypeMandarin
Size (English) Chest35.25 to 38.75 in.
Length (Metric)86.4 cm
Length (English) Sleeve33.75 in.
Length (Metric) Sleeve85.72 cm
Length (English)34 in.
Product TypeGeneral Purpose Frock
Seam TypeSerged
For Use With (Application)Lead and asbestos abatement and remedation, general maintenance and operations, spray painting, general clean-up
Unit SizeCase