▸
First Aid and Medical
▸
Patient Care Products
▸
Patient Care Simulators
▸
Water Rescue Manikins
Actual product may vary
Simulaids
Water Rescue Manikins
Used for water rescue trainings
Have Questions?
Catalog number
FIS19-819-211
Price (SGD)
-
Made of vinyl plastic with rust-resistant stainless steel skeletons, articulated joints
CPR capabilities are optional
Manikin will submerge to neck, when filled with water
Add additional 5 to 10 lb. (2.2 to 4.5kg) of weight to sink (not supplied)
Specifications
Product Type
Adult Water Rescue Manikin