KleenGuard™ V50 OTG™ Safety Glasses
Actual product may vary
Ansell
KleenGuard™ V50 OTG™ Safety Glasses
When you need to wear your prescription glasses and safety eye protection, you can trust KleenGuard OTG Over the Glasses Safety Glasses. They are made with a nylon frame for strength, durability and comfort and fit over most prescription glasses.
Have Questions?
Catalog NumberLens Tint
FIS19-818754Clear
Catalog number FIS19-818754
Price (SGD)
30.00
Each
Lens Tint:
Clear
Price (SGD)
30.00
Each
  • Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
  • See-through integrated side shields provide side protection without hindering peripheral vision
  • Hard coated for scratch resistance and longer life
  • Anti-fog coating for humid environment and temperature variances
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Certifications/ComplianceANSI Z87.1 and CSA Z94.3
Lens TintClear
Product TypeOTG safety spectacles
Frame ColorBlack
MaterialPolycarbonate
Frame MaterialPolycarbonate
UV ProtectionUV400
Unit SizeEach