Excel FR™ Lab Coats
Actual product may vary
VF
Excel FR™ Lab Coats
Protection: Arc Rating ATPV 7.7 calories/cm2
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-816-9532X-Large
FIS19-816-952X-Large
FIS19-816-756X-Small
Catalog number FIS19-816-953
Price (SGD)
235.00
Each
Size:
2X-Large
Price (SGD)
235.00
Each
Protection: Arc Rating ATPV 7.7 calories/cm2
  • Flame-resistant, 7 oz. 100% cotton
  • Button-front closure with lapel collar
  • Hemmed sleeve ends
  • Side-vent openings
  • Light blue
  • Hazard rating class 1
Specifications
Product TypeFlame-resistant Lab Coat
Size2X-Large
GenderUnisex
MaterialCotton
ColorLight Blue
Closure TypeButtons
DisposableReusable
SterilityNon-sterile
ResistanceFlame
Collar TypeLapel
ModelKEL2LBRG
Hazard RatingClass 1
Arc Flash Rating7.7 Calories/cm2
Certifications/ComplianceASTM F2302, NFPA 70E, ASTM F1506
For Use With (Application)Chemical, Oil, Gas, Petrochemical, Electrical, Utilities
IncludesSide Vent Openings
Weight (English)7 oz.
Weight (Metric)0.19 kg
Unit SizeEach