Reusable Pot Sink Aprons
Actual product may vary
PIP
Reusable Pot Sink Aprons
Suitable for food handling and preparation, general maintenance and chemical processing
Have Questions?
Catalog NumberLength (English)
FIS19-810-60836 in.
Catalog number FIS19-810-608
Price (SGD)
-
Length (English):
36 in.
  • 8 mil PVC/vinyl apron with neck ties
  • Comfortable to wear with superior flexibility
  • Excellent resistance to punctures and abrasion, chemicals, fats, oils and grease
  • Individually packaged
  • White
Specifications
TypeApron
SizeUniversal
MaterialVinyl
ColorWhite
DisposableReusable
Closure TypeTies
SterilityNon-sterile
Thickness8 mil
For Use With (Application)Great for food handling and preparation, industrial maintenance and chemical processing
Dimensions (L x W)35 x 36 in. (91.4 x 91.4 cm)
Length (English)36 in.
Width (English)35 in.
Seam TypeSeamless
Unit SizeDozen