▸
Facility Safety and Maintenance
▸
Emergency Eyewashes and Showers
▸
Eyewash and Shower Valves and Accessories
▸
Drench Shower Tester
Actual product may vary
Bradley
Drench Shower Tester
Provides a convenient way to comply with ANSI specifications for weekly testing on all drench showers without getting wet
Have Questions?
Catalog Number
Includes
FIS19-807-547
Funnel, Handle
Catalog number
FIS19-807-547
Price (SGD)
-
Includes:
Funnel, Handle
213.4cm (84 in.) chemically resistant nylon funnel with weighted bottom protects from splashing
Telescoping aluminum handle extends from 114.3cm (45 in.) to 182.9cm (72 in.)
Top diameter of 38.1cm (15 in.) and bottom diameter of 12.7cm (5 in.)
Yellow
Specifications
Includes
Funnel, Handle
Product Type
Drench Shower Tester