KleenGuard™ V90 Shield™ Safety Goggles with Face Shield
Actual product may vary
Ansell
KleenGuard™ V90 Shield™ Safety Goggles with Face Shield
When you have a job to tackle that involves flying particles or splashes, wearing safety glasses isn't enough. Protect yourself with the KleenGuard V90 Shield Safety Goggles with Face Shield.
Have Questions?
Catalog number FIS19-706-028
Price (SGD)
66.00
Each
Price (SGD)
66.00
Each
  • MonoGoggle XTR with full-face protection
  • Shield detaches from goggle
  • Blue frame with Visiclear Clear lens
  • Replacement shield available (see cat. no. 172831)
Specifications
Certifications/ComplianceANSI Z87.1-2010
Lens TintClear
Product TypeSafety Goggles
For Use With (Application)Provides full eye and face protection from liquids without primary eye protection underneath a face shield
Frame ColorBlue
MaterialPolycarbonate
UV ProtectionUV400
Unit SizeEach