BioClean™ Nerva BNAL Extra Long Nitrile Cleanroom Gloves
Actual product may vary
Ansell
BioClean™ Nerva BNAL Extra Long Nitrile Cleanroom Gloves
Nitrile 16 in. elbow length gloves are resistant to a range of chemicals and reduce the risk of cross contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-668-002Medium
FIS19-668-001Small
FIS19-668-003Large
Catalog number FIS19-668-002
Price (SGD)
2,203.00
Pack
Size:
Medium
Price (SGD)
2,203.00
Pack
  • Extra long 16 in. to provide elbow protection
  • Ambidextrous
  • Non-sterile
  • Textured, powder free, latex-free
  • ESD Properties
  • Processed in a cleanroom environment to ensure compatibility with ISO Class 4 operating conditions
  • Beaded cuff
  • Non-particulating easy tear packaging
  • Double-donnable
  • Ideal for Controlled and Critical Environments, Production and Manufacturing, Lab and R&D
Specifications
Certifications/Compliance (Europe)EN 420:2003 + A1:2009, EN 421:2010, EN 455 Part 2, EN ISO 374-1:2016, EN ISO 374-5:2016, Category III
Cleanroom ClassClass 10 (ISO 4)
Accelerator FreeYes
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)6.3 mil
Finger Thickness (Metric)0.16 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.9 mil
Palm Thickness (Metric)0.10 mm
PowderedPowder Free
Product TypeControlled Environment Gloves
SizeMedium
SterilityNon-sterile
Thickness (English)6.3 mil
Thickness (Metric)0.16 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBNAL-M
SeriesBioClean™ Nerva BNAL
Length (English)16 in.
Length (Metric)40 cm
Unit SizePack