STERI-PEROX™ Sterile Hydrogen Peroxide Solution
Actual product may vary
STERI-PEROX™ Sterile Hydrogen Peroxide Solution
STERI-PEROX™ Sterile Hydrogen Peroxide Solution
STERI-PEROX™ Sterile Hydrogen Peroxide Solution
STERI-PEROX™ Sterile Hydrogen Peroxide Solution
STERI-PEROX™ Sterile Hydrogen Peroxide Solution is designed for pharmaceutical and biotechnology operations.
Have Questions?
Catalog NumberVolume (English)Concentration
FIS19-547-13716 oz.3%
Catalog number FIS19-547-137
Price (SGD)
1,363.00
Case
Volume (English):
16 oz.
Concentration:
3%
Price (SGD)
1,363.00
Case
  • Ideal for use on walls, ceilings, floors and surfaces
  • Formulated with USP purified water and filtered at 0.2 μm
  • Completely traceable and validated for sterility and shelf life
Specifications
SterilitySterile
Certifications/ComplianceUSP, ISO
Concentration3%
FilteredFiltered
For Use With (Application)Sterilization of manufacturing, packaging and filling equipment in aseptic processes, for use in cleaning rotation cycles to maintain a clean environment
IncludesLot specific certificate of irradiation and certificate of analysis
Volume (English)16 oz.
Volume (Metric)473 mL
Unit SizeCase