Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Color | Material | Certifications/Compliance |
|---|---|---|---|---|
| FIS19-4 | 208.19 L | Black | Carbon Steel | UN A2/Y1.6/150, UN 1A2/x400/S-DOT/UN |
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.