GammaGuard CE™ Sterile Cleanroom Shoe Covers
Actual product may vary
GammaGuard CE™ Sterile Cleanroom Shoe Covers
GammaGuard CE™ Sterile Cleanroom Shoe Covers
International Enviroguard
GammaGuard CE™ Sterile Cleanroom Shoe Covers
Disposable sterile cleanroom shoe covers are made from high MVTR, innovative fabric. Each pair is individually packaged to help prevent cross-contamination.
Have Questions?
Catalog NumberSizeThickness
FIS19-472-642Standard31 mil
Catalog number FIS19-472-642
Price (SGD)
1,346.00
Case
Size:
Standard
Thickness:
31 mil
Price (SGD)
1,346.00
Case
Sterile shoe covers are made of comfortable fabric and include non-skid soles to help prevent accidental slips as you work. Shoe covers protect against liquids and particulates, so your shoes stay pristine. Each shoe cover is low-linting and sterilized to the highest Sterility Assurance Level (SAL) of 10-6. The low-linting fabric helps reduce the release of unwanted particulates in your work area while tunnelized elastic also contains particle shed. Each pair of shoe covers is individually bagged to help prevent cross-contamination. Each bag includes a red indicator dot for proof of sterilization.
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE™ helps keep wearers cooler vs many competitive fabrics
  • Tear-resistant fabric ensures your safety in demanding environments
  • Meets high manufacturing standards - made in an ISO-certified facility
  • Certificate of sterility included in each case
  • Bound seams
Specifications
SizeStandard
MaterialMicroporous Fabric
Latex FreeYes
ColorWhite
DisposableSingle-use
SterilitySterile
Closure TypeElastic Ankle
Sole TypeNon-skid
Seam TypeBound
Thickness31 mil
Certifications/ComplianceISO, ANSI ISEA 101-1996 R2034
Packaging Quantity200 Pairs/Cs.
Packaging TypeIndividually Packaged
Height (English)18 in.
Height (Metric)45.72 cm
Product TypeControlled Environments Shoe Covers
Unit SizeCase