Polyethylene Corrosives/Acid Cabinet
Actual product may vary
Polyethylene Corrosives/Acid Cabinet
Polyethylene Corrosives/Acid Cabinet
Polyethylene Corrosives/Acid Cabinet
Justrite
Polyethylene Corrosives/Acid Cabinet
Sturdy, completely metal-free poly cabinet for storing acids or bases without the worry of corrosion
Have Questions?
Catalog NumberDoor Style
FIS19-414-063Manual-close
Catalog number FIS19-414-063
Price (SGD)
5,212.00
Each
Door Style:
Manual-close
Price (SGD)
5,212.00
Each
Safely store strong acids such as hydrochloric, sulfuric or nitric acid; or bases such as sodium, potassium or calcium hydroxide in this durable, HDPE (high-density polyethylene) cabinet. With no metal parts to corrode, this cabinet holds up to 36, 2.5L (90L) bottles, and offers chemical resistance against spills or potential damaging vapors when storing acids or bases. The shelf adjusts on 3 in. (76-mm) increments, and has a load capacity of 125 lb. (57kg); choose from three positions to optimize container storage. A removable yet sturdy sump cover, which also serves as a bottom shelf, conceals the fully integrated, leakproof sump with an 8.5-gal (32L) capacity. Recessed channels in shelf and base help direct spills to the back of the cabinet and down into the sump. Three application-specific labels identify contents to safely store and segregate incompatible chemicals — ACID, BASE, and CORROSIVE. Doors open nearly 180 degrees for easy access, engage firmly with the body when closed, and accept a padlock (not included) for added security.
Specifications
Depth (English) Interior18 in.
Depth (Metric) Interior457 mm
Door StyleManual-close
For Use With (Application)Store acids or bases
Height (English) Interior28 in.
Height (Metric) Exterior889 mm
Height (Metric) Interior711 mm
Load Bearing Capacity36 x 2.5 L Bottles
No. of Shelves2
Width (English) Interior31.25 in.
Width (Metric) Exterior914 mm
Width (Metric) Interior794 mm
ColorBlue
Height (English) Exterior35 in.
MaterialHDPE
Width (English) Exterior36 in.
Unit SizeEach