Disposal Can Spill Basin
Actual product may vary
Disposal Can Spill Basin
Disposal Can Spill Basin
Disposal Can Spill Basin
Justrite
Disposal Can Spill Basin
Capture inadvertent leakage. The Disposal Can Spill Basin holds containers up to 30.5cm (12 in.) diameter.
Have Questions?
Catalog NumberCapacity (English)
FIS19-413-8245 gal.
Catalog number FIS19-413-824
Price (SGD)
479.00
Each
Capacity (English):
5 gal.
Price (SGD)
479.00
Each
Accepts disposal cans with a diameter of 30.5cm (12 in.). Recommended for holding up to 18.9L (5 gal.).
  • Leakproof
  • Translucent
  • Durable
  • Polyethylene
Specifications
Certifications/ComplianceEPA
ColorTranslucent
Capacity (English)5 gal.
Capacity (Metric)18.9 L
MaterialPolyethylene
TypeSpill Basin
For Use With (Application)Catching accidental spills when filling safety disposal can
ShapeSquare
Length (English)14.875 in.
Length (Metric)37.9 cm
Width (English)14.875 in.
Width (Metric)37.9 cm
Height (English)7.625 in.
Height (Metric)19.4 cm
Unit SizeEach