Ultra-Hard Top P4 Plus™ Containment Unit
Ultra-Hard Top P4 Plus™ Containment Unit
Actual product may vary
Youngstown Barrel and Drum
Ultra-Hard Top P4 Plus™ Containment Unit
Safely store four drums outdoors
Have Questions?
Catalog NumberMaterial
FIS19-339-030Polyethylene
Catalog number FIS19-339-030
Price (SGD)
-
Material:
Polyethylene
  • Unique “dual-closure” offers easy access from either side
  • “Roll-top” covers easily lift from waist height to access drum tops
  • Low-profile (8.75 in.) containment pallet positions drums at a safe, convenient level to pour wastes; with drain
  • Forklift compatible and lockable, with an all-polyethylene construction that will not rust or corrode
  • Optional loading ramp (cat. no. 19-130-4953) available; stores inside of containment ramp when not in use
Specifications
Certifications/Compliance40 CFR 264.175
MaterialPolyethylene
TypeMulti-drum Containment
For Use With (Application)Safely storing four drums outdoors
Load Bearing Capacity (English)9000 lb.
Load Bearing Capacity (Metric)4082 kg
Weight (English)440 lb.
Weight (Metric)200 kg
Unit SizeEach