PharmaMOP™ Cleanroom Mop Heads, Sterile
Actual product may vary
Foamtec
PharmaMOP™ Cleanroom Mop Heads, Sterile
For cleaning and disinfection of rough surfaces and high soil areas
Have Questions?
Catalog NumberMaterialProduct Type
FIS19-304-822PolyesterHigh Fluid Capacity Cleanroom Mop Head
Catalog number FIS19-304-822
Price (SGD)
2,543.00
Case
Material:
Polyester
Product Type:
High Fluid Capacity Cleanroom Mop Head
Price (SGD)
2,543.00
Case
Enables the quick, efficient cleaning of surfaces stained with disinfectant residues
  • Gamma irradiated
  • EzGLIDE™ polyester mop head
  • 0.125 in. foam core
Specifications
Product TypeHigh Fluid Capacity Cleanroom Mop Head
SterilitySterile
AutoclavableAutoclavable
Size1.45 lb.
Quantity1.45 lb.
For Use With (Application)For cleaning and disinfection of rough surfaces and high soil areas, ISO class 1 to 7 cleanrooms
Certifications/ComplianceISO 9001:2000
MaterialPolyester
Unit SizeCase