MiraSWAB™ 1500 Series Cleanroom Swabs
MiraSWAB™ 1500 Series Cleanroom Swabs
Actual product may vary
Foamtec
MiraSWAB™ 1500 Series Cleanroom Swabs
Offers cleaning performance of micro fiber fabrics with scratch-free properties of foam for Optics, HDD, Semiconductor and Life Science applications
Have Questions?
Catalog NumberLength (Metric)Tip StyleShapeWidth (Metric) Tip
FIS19-304-630152.4 mmFlexibleCylinder1.3 mm
Catalog number FIS19-304-630
Price (SGD)
-
Length (Metric):
152.4 mm
Tip Style:
Flexible
Shape:
Cylinder
Width (Metric) Tip:
1.3 mm
Unique fabric over foam and fabric over fabric construction enables easy cleaning of scratch sensitive surfaces and blind areas as the two layers provide a soft conformable tool resulting in a swab that is soft, yet both resistant to abrasion and solvents.
  • Fabric over foam construction compatible with aggressive solvents such as acetone, Kyzen and MEK
  • Super soft foam core enables the swabs to conform to articulated surfaces for good cleaning efficiency
  • Micro fiber fabric enables excellent removal of difficult to remove stains and particles
  • Proprietary cleaning process ensures low extractable for streak free cleaning
Specifications
Diameter (English) Handle0.11 in.
Length (English)6 in.
For Use With (Application)The cleaning performance of micro Fiber Fabrics with the scratch-free properties of Foam for optics, HDD, semiconductor and life science applications
Handle ColorBlue
Product TypeSwab
Diameter (Metric) Handle2.8 mm
Length (Metric)152.4 mm
Length (English) Handle5.26 in.
Length (Metric) Handle133.6 mm
ShapeCylinder
SizeMedium
Thickness (Metric)17.8 mm
Length (English) Tip0.7 in.
Length (Metric) Tip17.8 mm
Width (Metric) Tip1.3 mm
Width (English) Tip0.1 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip StyleFlexible
Tip MaterialMicrofiber
Unit SizePack