Concealed Snap Front Cotton Lab Coats
Actual product may vary
VF
Concealed Snap Front Cotton Lab Coats
Flame-resistant coats ideal for manufacturing industry or in university labs.
Have Questions?
Catalog NumberSizeSize (English) Chest
FIS19-296-5852X-Large52 to 54 in.
Catalog number FIS19-296-585
Price (SGD)
289.00
Each
Size:
2X-Large
Size (English) Chest:
52 to 54 in.
Price (SGD)
289.00
Each
Flame-resistant coats ideal for manufacturing industry or in university labs
  • Concealed snap front
  • Lapel collar
  • Hemmed sleeve ends
  • Side vent openings
  • Flame-resistant
  • Fabric composition: 88% Cotton/12% Nylon
  • One chest and two hip pockets
Specifications
Product TypeFlame-resistant Lab Coat
Size2X-Large
GenderMen's
MaterialCotton, Nylon
ColorNavy
Closure TypeSnaps
Cuff StyleHemmed
DisposableReusable
SterilityNon-sterile
ResistanceFlame
StyleRegular
No. of Pockets3
Collar TypeLapel
FinishAmmonia Cure, Heat Cure
ModelKLL6NV RG
Size (English) Chest52 to 54 in.
Certifications/ComplianceNFPA 2112
Unit SizeEach