Kroflex 840 Cut-Resistant Gloves, Latex Coated
Kroflex 840 Cut-Resistant Gloves, Latex Coated
Actual product may vary
MAPA
Kroflex 840 Cut-Resistant Gloves, Latex Coated
Enhanced non-slip feature offers ultra-protection when handling heavy/sharp objects in wet environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-249-7579
FIS19-249-7557
Catalog number FIS19-249-757
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
9
Ideal for use when handling glass sheets, hot plastic parts, composite materials
  • Natural rubber crinkle palm coating
  • Mechanical durability
  • Anatomical shape and flexibility
  • Comfortable for extended wear, with a secure grip
Specifications
ANSI Abrasion RatingANSI 2
Certifications/Compliance (Europe)EN 421 , EN 407 , EN 511 , CE 3543
Coating MaterialLatex
ColorBlue, Gray
Cut Resistance RatingANSI A4
Latex FreeNo
MaterialHDPE
Product TypeCut-resistant Gloves
Size9
EN388 StandardEN 388
For Use With (Application)Handling Glass, hot Plastic parts and waste treatment applications
Glove Cuff StyleKnit
Model840419
SeriesKroflex 840
Length (English)10.5 in.
Length (Metric)26.5 cm
Unit SizePAA