Butoflex 651 Short Wear Chemical Resistant Butyl Gloves
Actual product may vary
MAPA
Butoflex 651 Short Wear Chemical Resistant Butyl Gloves
Unlined butyl gloves feature an embossed exterior finish for a secure grip on slippery objects. Ideal for chemical, mechanical and pharmaceutical applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-249-7507
FIS19-249-7518
Catalog number FIS19-249-750
Price (SGD)
167.00
Pair
Size:
7
Price (SGD)
167.00
Pair
  • Butyl protection for chemical gases and liquids, such as ketones, esters and amines
  • Ideal for use with paints, resins, varnish and inks
  • Optimal embossed textured grip
  • Comfortable, unlined medium weight glove
  • Rolled cuff to catch drips and facilitate attachment to chemical suits
Specifications
Certifications/ComplianceCE EUR CAT 3, EN 388 0111
Certifications/Compliance (Europe)EN 16350, Cat 3 CE 0334
ApplicationsIndustrial
ColorBlack
Latex FreeYes
MaterialButyl
Product TypeButyl Gloves
Size7
Thickness (English)20 mil
Thickness (Metric)0.50 mm
EN388 StandardEN 388:0010X
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESABCILMNOS
EN ISO 374 - 5Bacteria, Fungi only
For Use With (Application)Chemical gases and liquids such as ketones, esters and amines
Glove Cuff StyleRolled
Grip DesignEmbossed
LiningUnlined
Model651317
Length (English)14 in.
Length (Metric)35.5 cm
Unit SizePair